Learn More
Mopec™ Circular Specimen Chuck for Leica, TBS, and Tanner Cryostats, 25 mm Kit
SDP
Catalog No. 22445787 Shop All Mopec Products
Change view
Click to view available options
For Use With (Equipment):
Leica, TBS, and Tanner Cryostats
Catalog No. For Use With (Equipment)
22-445-787 Leica, TBS, and Tanner Cryostats
1 options
Catalog No. 22-445-787 Supplier Mopec™ Supplier No. SP102KT
Add to Cart
Add to Cart
Round, aluminum specimen chucks suitable for Leica (except CM1950 model), TBS, and Tanner Cryostats.
  • Includes 6 x 25 mm, 2 x 30 mm, and 2 x 40 mm chucks in different colors, an angular brush, and a teasing needle
Specifications
For Use With (Equipment) Leica, TBS, and Tanner Cryostats