Learn More
Mopec™ Circular Specimen Chuck for Leica, TBS, and Tanner Cryostats, 30 mm Kit
SDP
Catalog No. 22445789 Shop All Mopec Products
Change view
Click to view available options
For Use With (Equipment):
Leica, TBS, and Tanner Cryostats
Catalog No. For Use With (Equipment)
22-445-789 Leica, TBS, and Tanner Cryostats
1 options
Catalog No. 22-445-789 Supplier Mopec™ Supplier No. SP103KT
Add to Cart
Add to Cart
Round, aluminum specimen chucks suitable for Leica (except CM1950 model), TBS, and Tanner Cryostats.
  • Includes 2 x 25 mm, 6 x 30 mm, and 2 x 40 mm in different colors, an angular brush, and a teasing needle
Specifications
For Use With (Equipment) Leica, TBS, and Tanner Cryostats