Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ Circular Specimen Chucks for SHANDON Cryostats
SDP

Catalog No. 22445770 Shop All Mopec Products
Change view
Click to view available options
Color:
Green
Orange
Yellow
Length (Metric):
30 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Length (Metric)
22-445-770 Yellow 30 mm
22-445-558 Green 30 mm
22-445-361 Orange 30 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-445-770 Supplier Mopec™ Supplier No. SP143YE
Add to Cart
Add to Cart

Available in multiple sizes and colors for use with SHANDON Cryostats.

  • Round, aluminum specimen chucks

Specifications

For Use With (Equipment) SHANDON Cryostats
Length (Metric) 30 mm
Color Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.