Learn More
Cole-Parmer CIRCULATING BATH,7L,120VAC
Supplier:  Cole-Parmer 2087297
Encompass_Preferred
Cole-Parmer Polystat Programmable Heated Circulating Bath, 7 L; 120 VAC. Advanced programmability to suit your complex needs...preciselyIntuitive touch-screen display includes multiple selectable home screensTemperature trends can easily be viewed for up to 10 daysSchedule time/temperature events and tests without restrictions on number of programs or stepsTimer can be set from 0 to 999 minutes in 1-minute increments
Catalog No. NC1938852