Learn More
Polyscience 7L Heated Circulator, Standard Digital Controller (Ambient +10 to 170°C); 120V, 60Hz
SDP
Catalog No. NC1032944
Encompass_Preferred
PolyScience™ 7 Liter Heated CirculatorStandard Digital ControllerAmbient +10 to 170°C Working Temperature RangeTemperature Stability: ±0.04°C1100W HeaterMaximum Pressure: 3.5 psi (0.24 bar) Maximum Pressure Flow Rate: 2.9 gpm (11 LPM) 120V, 60HzFeatures:• Large, universal icon and English display• Swivel 180™ Rotating Controller• On-board connectivity: RS-232 serial output• On-screen prompts• Automatic performance optimization• Single-point calibration capability
Catalog No. NC1032944 Supplier Polyscience Supplier No. SD07H170A11B
PolyScience™ 7 Liter Heated CirculatorStandard Digital ControllerAmbient +10 to 170°C Working Temperature RangeTemperature Stability: ±0.04°C1100W HeaterMaximum Pressure: 3.5 psi (0.24 bar) Maximum Pressure Flow Rate: 2.9 gpm (11 LPM) 120V, 60HzFeatures:• Large, universal icon and English display• Swivel 180™ Rotating Controller• On-board connectivity: RS-232 serial output• On-screen prompts• Automatic performance optimization• Single-point calibration capability