Learn More
Cole-Parmer Techne TU-20D Advanced Digital Immersion Circulator; 120 V, 50/60 Hz
Catalog No. 501960466 Shop All Cole-Parmer Products
Encompass
This digital immersion circulator offers a wide temperature range of 40 to 200°C (with optional chiller). Circulator includes all of the features found on the basic digital circulator model, plus RS-232 interface capability. Bidirectional RS-232 lets you control bath remotely and download data to a computer for further analysis.
Catalog No. 50-196-0466 Supplier Cole-Parmer Supplier No. 0126213
This digital immersion circulator offers a wide temperature range of 40 to 200°C (with optional chiller). Circulator includes all of the features found on the basic digital circulator model, plus RS-232 interface capability. Bidirectional RS-232 lets you control bath remotely and download data to a computer for further analysis.