Learn More
Cole-Parmer Techne TE-10D High-Stability Digital Immersion Circulator; 240V, 50/60Hz
Catalog No. 501960465 Shop All Cole-Parmer Products
Encompass
This immersion circulator offers precise PID temperature control at a competitive price. Circulator features digital LED display, ±0.01°C stability, overtemperature indicator light, pressure pump, and low-liquid level cutoff.
Catalog No. 50-196-0465 Supplier Cole-Parmer Supplier No. 0126207
This immersion circulator offers precise PID temperature control at a competitive price. Circulator features digital LED display, ±0.01°C stability, overtemperature indicator light, pressure pump, and low-liquid level cutoff.