Learn More
Cole-Parmer IC-400 Standard Immersion Circulator; 120 VAC
Catalog No. NC2127890 Shop All Cole-Parmer Products
Encompass_Preferred
Smartly suited for lab and industrial use, this immersion circulator has a large backlit digital display, yet a slim overall profile requiring minimal reservoir space. From across the room, users can readily see the process value and set value, as well as heating and circulating icons. Four buttons enable easy setting of on/off/standby, point value, °F/°C readings and offset calibration value; min/max temperatures can also be easily selected. A protective housing prevents direct contact with the integral pump and heater coil yet is easy to clean and inspect. The adjustable mounting clamp attaches securely to flat and rounded tank walls.
This item is not returnable. View return policy
Catalog No. NC2127890 Supplier Cole-Parmer Supplier No. 2087373
Smartly suited for lab and industrial use, this immersion circulator has a large backlit digital display, yet a slim overall profile requiring minimal reservoir space. From across the room, users can readily see the process value and set value, as well as heating and circulating icons. Four buttons enable easy setting of on/off/standby, point value, °F/°C readings and offset calibration value; min/max temperatures can also be easily selected. A protective housing prevents direct contact with the integral pump and heater coil yet is easy to clean and inspect. The adjustable mounting clamp attaches securely to flat and rounded tank walls.