Learn More
Polyscience 28L Refrigerated Circulator, Standard Digital Controller (-30 to 170°C); 120V, 60Hz
SDP
Catalog No. NC1753875
Encompass_Preferred
PolyScience™ 28 Liter Refrigerated CirculatorStandard Digital Controller-30 to 170°C Working Temperature RangeTemperature Stability: ±0.04°CCooling Capacity at 20°C: 915W 1100W HeaterFlow Rate: 2.9 gpm (11 LPM)Maximum Pressure: 3.5 psi (0.24 bar)120V, 60HzFeatures:• Large, universal icon and English display• Swivel 180™ Rotating Controller• On-board connectivity: RS-232 serial output• On-screen prompts• Automatic performance optimization• Single-point calibration capability
Catalog No. NC1753875 Supplier Polyscience Supplier No. SD28R30A11B
Add to Cart
Add to Cart
Request a Quote
PolyScience™ 28 Liter Refrigerated CirculatorStandard Digital Controller-30 to 170°C Working Temperature RangeTemperature Stability: ±0.04°CCooling Capacity at 20°C: 915W 1100W HeaterFlow Rate: 2.9 gpm (11 LPM)Maximum Pressure: 3.5 psi (0.24 bar)120V, 60HzFeatures:• Large, universal icon and English display• Swivel 180™ Rotating Controller• On-board connectivity: RS-232 serial output• On-screen prompts• Automatic performance optimization• Single-point calibration capability