Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Armstrong Ceiling Tile, Mineral Fiber, 24 x 24 in, Angled Tegular

Catalog No. 502838224
Encompass

This ceiling tile is a 24 x 24 inch panel designed with an angled tegular edge. Made from wet-formed mineral fiber, it offers good acoustic properties with a Noise Reduction Coefficient (NRC) of 0.70 and a Ceiling Attenuation Class (CAC) of 35, making it suitable for various commercial applications. It also features high light reflectance and resistance to sag, humidity, mold, and mildew.

  • Class A fire performance.
  • Disinfectable following CDC recommendations.
  • Contains 71% recycled content and is recyclable.
  • Made in U.S.A. and compliant with Build America, Buy America Act.
  • Installation method: grid (suspended).

Catalog No. 50-283-8224 Supplier MSC Supplier No. 85082097
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This ceiling tile is a 24 x 24 inch panel designed with an angled tegular edge. Made from wet-formed mineral fiber, it offers good acoustic properties with a Noise Reduction Coefficient (NRC) of 0.70 and a Ceiling Attenuation Class (CAC) of 35, making it suitable for various commercial applications. It also features high light reflectance and resistance to sag, humidity, mold, and mildew.

  • Class A fire performance.
  • Disinfectable following CDC recommendations.
  • Contains 71% recycled content and is recyclable.
  • Made in U.S.A. and compliant with Build America, Buy America Act.
  • Installation method: grid (suspended).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.