Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cirsimarin | 13020-19-4 | 1 MG
SDP

Catalog No. 5000419329
Encompass_Preferred

Cirsimarin is a potent antilipogenic flavonoid isolated from Microtea debilis. It exerts a potent antilipogenic effect, decreasing adipose tissue deposition in mice. Its lipolytic activity results from both its antagonist activity on the adenosine A1 receptor and its inhibitory effect on phosphodiesterase, making it suitable for various research applications.

  • Potent antilipogenic flavonoid
  • Decreases adipose tissue deposition in mice
  • Antagonist of adenosine A1 receptor
  • Inhibitor of phosphodiesterase
  • Purity: 99.71%
  • Molecular Weight: 476.43
  • Formula: C23H24O11
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Storage at -20°C, protected from light
  • Soluble in DMSO at 50 mg/mL

Catalog No. 50-004-19329 Supplier Medchemexpress LLC Supplier No. HYN72721MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cirsimarin is a potent antilipogenic flavonoid isolated from Microtea debilis. It exerts a potent antilipogenic effect, decreasing adipose tissue deposition in mice. Its lipolytic activity results from both its antagonist activity on the adenosine A1 receptor and its inhibitory effect on phosphodiesterase, making it suitable for various research applications.

  • Potent antilipogenic flavonoid
  • Decreases adipose tissue deposition in mice
  • Antagonist of adenosine A1 receptor
  • Inhibitor of phosphodiesterase
  • Purity: 99.71%
  • Molecular Weight: 476.43
  • Formula: C23H24O11
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Storage at -20°C, protected from light
  • Soluble in DMSO at 50 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.