Learn More
Selleck Chemical LLC Cirtuvivint E2668-5MG
SDP
Catalog No. 5000039940
Encompass_Preferred
Cirtuvivint (SM08502) is a potent and orally active inhibitor of CDC-like kinase (CLK) and a DYRK (dual-specificity tyrosine kinase) that targets mRNA splicing and is optimized for Wnt pathway inhibition Cirtuvivint inhibits serine and arginine-rich splicing factor (SRSF) phosphorylation and disrupts spliceosome activity Cirtuvivint can be used for solid tumor research
Catalog No. 50-000-39940 Supplier Selleck Chemical LLC Supplier No. E26685MG
Cirtuvivint (SM08502) is a potent and orally active inhibitor of CDC-like kinase (CLK) and a DYRK (dual-specificity tyrosine kinase) that targets mRNA splicing and is optimized for Wnt pathway inhibition Cirtuvivint inhibits serine and arginine-rich splicing factor (SRSF) phosphorylation and disrupts spliceosome activity Cirtuvivint can be used for solid tumor research