Learn More
Medchemexpress LLC HY-137435 5mg , Cirtuvivint CAS:2143917-62-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1374355MG
Encompass_Preferred
HY-137435 5mg Cirtuvivint CAS: 2143917-62-6 Cirtuvivint (SM08502) is a potent and orally active CDC-like kinase (CLK) inhibitor. Cirtuvivint can be used for solid tumors research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9249