Learn More
Strem, An Ascensus Company CAS# 983-80-2. 1g. cis-1,2-Bis(diphenylphosphino)ethylene, min. 98%. MFCD00003046
SDP
Supplier:  Strem, An Ascensus Company 1502501G
Encompass
CAS# 983-80-2. 1g. cis-1,2-Bis(diphenylphosphino)ethylene, min. 98%. MFCD00003046. Molecular Weight: 396.41. Molecular Formula: cis-(C6H5)2PCH=CHP(C6H5)2. Color/form: white xtl. Strem# 15-0250. http://www.strem.com/catalog/v/15-0250/
Catalog No. 50-138-5362