Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-1,2-Dichloroethene, MilliporeSigma™ Supelco™

Catalog No. 111020127 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0127 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0127 Supplier MilliporeSigma Supelco Supplier No. 48597
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 156-59-2
Density 1.284 g/mL (at 25°C)
Flash Point 6°C
Physical Form Neat
Linear Formula ClCH=CHCl
MDL Number MFCD00000925
Melting Point -80°C
Boiling Point 60°C
Synonym cis-1,2-Dichloroethylene; cis-Acetylene dichloride
Type Organic Pollutant Standard
Quantity 1000 mg
Formula Weight 96.94
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.