Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich cis-1,2-Dichloroethylene

Catalog No. D6200450G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D6200450G 50 g
D6200410G 10 g
D620041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D6200450G Supplier Sigma Aldrich Supplier No. D6200450G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

cis-1,2-Dichloroethylene

Specifications

Melting Point -80°C (lit.)
CAS 156-59-2
Density 1.284 g/mL (at 25°C (literature))
Quantity 50 g
Refractive Index n20/D 1.449 (literature)
Molecular Formula C2H2Cl2
Linear Formula ClCH=CHCl
MDL Number MFCD00000925
Synonym cis-1,2-DCE; cis-Acetylene dichloride
Boiling Point 60°C (lit.)
Molecular Weight (g/mol) 96.94
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.