Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-1,4-Cyclohexanediamine Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. C17995G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17995G 5 g
C179925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17995G Supplier TCI America Supplier No. C17995G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2121-79-1
Molecular Formula C6H16Cl2N2
Molecular Weight (g/mol) 187.108
MDL Number MFCD20487918
InChI Key LSWJXVMHLYPGPM-UHFFFAOYSA-N
Synonym cis-1,4-Diaminocyclohexane Dihydrochloride
PubChem CID 67930328
IUPAC Name cyclohexane-1,4-diamine;dihydrochloride
SMILES C1CC(CCC1N)N.Cl.Cl

Specifications

Color White
Quantity 5 g
Formula Weight 187.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material cis-1,4-Cyclohexanediamine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.