Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich cis-1,5-Cyclooctanediol

Catalog No. 17903550G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-903-550G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-903-550G Supplier Sigma Aldrich Supplier No. 17903550G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

cis-1,5-Cyclooctanediol

Specifications

CAS 23418-82-8
Melting Point 73°C to 75°C (lit.)
Boiling Point 105°C to 107°C (0.05 mmHg)
Molecular Formula C8H16O2
Linear Formula C8H14(OH)2
MDL Number MFCD00001746
Quantity 250 mg
Molecular Weight (g/mol) 144.21
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.