Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-1,8-p-Menthanediol monohydrate, 98%, Thermo Scientific™

Catalog No. AAA1701114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1701114 25 g
AAA1701122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1701114 Supplier Thermo Scientific Chemicals Supplier No. A1701114

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2451-01-6
MDL Number MFCD00167312

Specifications

Melting Point 114°C to 117°C
Molecular Formula C10H22O3
Quantity 25 g
Synonym terpin hydrate, p-menthane-1,8-diol monohydrate, terpin monohydrate, terpin cis-form hydrate, 4-2-hydroxypropan-2-yl-1-methylcyclohexanol hydrate, terpin hydrate usp, terpin hydrate cis-form, cyclohexanemethanol, 4-hydroxy-alpha,alpha,4-trimethyl-, monohydrate
InChI Key JGKJMBOJWVAMIJ-UHFFFAOYSA-N
SMILES CC1(CCC(CC1)C(C)(C)O)O.O
IUPAC Name 4-(2-hydroxypropan-2-yl)-1-methylcyclohexan-1-ol;hydrate
Molecular Weight (g/mol) 190.28
PubChem CID 17141
Formula Weight 190.28 (172.27 Anhydrous)
Percent Purity 98%
Chemical Name or Material cis-1,8-p-Menthanediol monohydrate
Show More Show Less
EINECSNumber 209-279-5
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.