Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
cis-10-Heptadecenoic acid
Spécifications
Spécifications
| CAS | 29743-97-3 |
| Molecular Formula | C17H32O2 |
| Linear Formula | C17H32O2 |
| MDL Number | MFCD00133175 |
| Quantity | 1 g |
| Molecular Weight (g/mol) | 268.43 |
| Percent Purity | ≥99% |
| Biological Activity | Respiratory System |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'