Learn More
Cayman Chemical cis-10-hydroxy Nortriptyline
SDP
Catalog No. 502913606
Encompass_Preferred
An active metabolite of amitriptyline and nortriptyline; formed from amitriptyline and nortriptyline by CYP2D6; inhibits norepinephrine reuptake in rat cortical slices (EC50 = 160 nM)
Catalog No. 50-291-3606 Supplier Cayman Chemical Supplier No. 36659500 UG
An active metabolite of amitriptyline and nortriptyline; formed from amitriptyline and nortriptyline by CYP2D6; inhibits norepinephrine reuptake in rat cortical slices (EC50 = 160 nM)