Learn More
Cayman Chemical cis-10-Nonadecenoic Acid, Purity: ≥98%, CAS: 73033-09-7, 250 mg
SDP
Catalog No. 502984452
Encompass_Preferred
A monounsaturated long chain fatty acid with potential antitumor and p53 inhibitory activity
Catalog No. 50-298-4452 Supplier Cayman Chemical Supplier No. 19749250 MG
A monounsaturated long chain fatty acid with potential antitumor and p53 inhibitory activity