Learn More
Cayman Chemical cIs-10-NonadecenoIc AcId 500mg
SDP
Catalog No. 502984709
Encompass_Preferred
A monounsaturated long chain fatty acid with potential antitumor and p53 inhibitory activity
Catalog No. 50-298-4709 Supplier Cayman Chemical Supplier No. 19749500 MG
A monounsaturated long chain fatty acid with potential antitumor and p53 inhibitory activity