Learn More
Medchemexpress LLC cis-11-Hexadecenal 500mg
SDP
Catalog No. 5000204899
Encompass_Preferred
cis-11-Hexadecenal has a fatty waxy odor and is commonly used as a flavoring agent in the food and beverage industry In addition it can be used as an intermediate in the synthesis of various organic compounds including fragrances pharmaceuticals and agrochemicals Its unique chemical properties make it an important ingredient in a variety of industrial processes notably in the production of perfumes and air fresheners
Catalog No. 50-002-04899 Supplier Medchemexpress LLC Supplier No. HYW127507500MG
cis-11-Hexadecenal has a fatty waxy odor and is commonly used as a flavoring agent in the food and beverage industry In addition it can be used as an intermediate in the synthesis of various organic compounds including fragrances pharmaceuticals and agrochemicals Its unique chemical properties make it an important ingredient in a variety of industrial processes notably in the production of perfumes and air fresheners