Learn More
Abcam cis-11-Methyl-2-dodecenoic acid, Diffusible signal factor, 100MG
Catalog No. 502705346
Encompass_Preferred
MW 212.33 g/mol, Purity >97%. Alpha, beta unsaturated fatty acid. Diffusible signal factor (DSF) in extracellular microbial and fungal communication systems. Cell-cell communication signal involved in the synchronization of virulence gene expression and biofilm dispersal of the pathogen Xanthomonas campestris pv. campestris (Xcc).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5346 Supplier Abcam Supplier No. AB145274100MG
MW 212.33 g/mol, Purity >97%. Alpha, beta unsaturated fatty acid. Diffusible signal factor (DSF) in extracellular microbial and fungal communication systems. Cell-cell communication signal involved in the synchronization of virulence gene expression and biofilm dispersal of the pathogen Xanthomonas campestris pv. campestris (Xcc).
The product is subject to the following: Abcam Restricted Use Statement