Learn More
Apexbio Technology LLC cis-11-Octadecenoic acid 506-17-2 100mg
SDP
Catalog No. 5000095989
Encompass_Preferred
cis-11-Octadecenoic acid (CAS 506-17-2) is a monounsaturated fatty acid of the -7 family containing an 18-carbon backbone with a cis double bond at the 11th carbon Present in various plant and animal lipids this compound contributes to the structural integrity and fluidity of cellular membranes by integrating into phospholipid bilayers It is implicated in modulating membrane permeability and participates in metabolic processes related to lipid homeostasis Studies also indicate potential roles in regulating inflammatory pathways and supporting cardiovascular function making it relevant for research on lipid metabolism and disease mechanism exploration
Catalog No. 50-000-95989 Supplier Apexbio Technology LLC Supplier No. A9964100
cis-11-Octadecenoic acid (CAS 506-17-2) is a monounsaturated fatty acid of the -7 family containing an 18-carbon backbone with a cis double bond at the 11th carbon Present in various plant and animal lipids this compound contributes to the structural integrity and fluidity of cellular membranes by integrating into phospholipid bilayers It is implicated in modulating membrane permeability and participates in metabolic processes related to lipid homeostasis Studies also indicate potential roles in regulating inflammatory pathways and supporting cardiovascular function making it relevant for research on lipid metabolism and disease mechanism exploration