Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-13-Docosenol 98.0+%, TCI America™
SDP

Catalog No. D21745G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21745G 5 g
D21741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21745G Supplier TCI America Supplier No. D21745G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 629-98-1
Molecular Formula C22H44O
Molecular Weight (g/mol) 324.59
MDL Number MFCD00055936
InChI Key CFOQKXQWGLAKSK-KTKRTIGZSA-N
Synonym Erucyl Alcohol
PubChem CID 5354168
ChEBI CHEBI:77416
IUPAC Name (13Z)-docos-13-en-1-ol
SMILES CCCCCCCC\C=C/CCCCCCCCCCCCO

Specifications

Melting Point 33°C
Color White-Yellow
Boiling Point 225°C
Quantity 5 g
Formula Weight 324.59
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material cis-13-Docosenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.