Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-2-Amino-1-cyclohexanecarboxylic acid, 98%, Thermo Scientific™

Catalog No. AC265620010
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC265620010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC265620010 Supplier Thermo Scientific Chemicals Supplier No. 265620010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5691-20-3
Assay Percent Range 98%
Molecular Formula C7H13NO2
MDL Number MFCD00143980,MFCD03790919
Synonym cis-2-aminocyclohexanecarboxylic acid, cis-2-amino-1-cyclohexanecarboxylic acid, 1r,2s-2-aminocyclohexanecarboxylic acid, 1r,2s-2-aminocyclohexane-1-carboxylic acid, 2-aminocyclohexanecarboxylic acid #, 1s,2r/1r,2s-2-aminocyclohexanecarboxylic acid, cis-2-amino-cyclohexanecarboxylic acid, cis-2-amino-1-cyclohexane carboxylic acid, cis-2-amino-1-cyclohexane-carboxylic acid, cyclohexanecarboxylic acid, 2-amino-, cis
InChI Key USQHEVWOPJDAAX-RITPCOANSA-N
SMILES [NH3+][C@H]1CCCC[C@H]1C([O-])=O
IUPAC Name (1R,2S)-2-azaniumylcyclohexane-1-carboxylate
Molecular Weight (g/mol) 143.19
PubChem CID 2724639
Formula Weight 143.19
Percent Purity ≥97.5% (on dry substance)
Chemical Name or Material cis-2-Amino-1-cyclohexanecarboxylic acid, 98%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.