Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-2-Amino-1-cyclopentanecarboxamide, 98%

Catalog No. AC334420010
Click to view available options
Quantity:
1 g

Catalog No. AC334420010

Supplier: Thermo Scientific Chemicals 334420010

Only null left
Add to Cart

CAS: 135053-11-1 | C6H12N2O | 128.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135053-11-1
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.17
MDL Number MFCD01863253
InChI Key FUGFTUCRJJFPES-UHNVWZDZSA-N
Synonym cis-2-aminocyclopentanecarboxamide, cis-2-amino-1-cyclopentanecarboxamide, 1r,2s-2-aminocyclopentane-1-carboxamide, 1r,2s-2-aminocyclopentanecarboxamide, 2beta-aminocyclopentane-1beta-carboxamide, +/--cis-2-aminocyclopentanecarboxamide, cis-2-amino-cyclopentanecarboxylic acid amide, cyclopentanecarboxamide, 2-amino-, 1r,2s, 1r,2s-2-amino-cyclopentanecarboxylic acid amide, cyclopentanecarboxamide, 2-amino-, 1r,2s-9ci
PubChem CID 2733686
IUPAC Name (1R,2S)-2-aminocyclopentane-1-carboxamide
SMILES C1CC(C(C1)N)C(=O)N

Specifications

Melting Point 131.0°C to 137.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula C5H8(NH2)CONH2
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol,chloroform,ethanol,dmso,,dmf,hydrochloric acid
Quantity 1 g
Formula Weight 128.17
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material cis-2-Amino-1-cyclopentanecarboxamide, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.