Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cis-2-butene-1,4-diol | 6117-80-2 | MFCD00002924 | 96.0% | 88.11 g/mol | C4H8O2 | 500 MG
SDP

Catalog No. 5000301561
Encompass_Preferred

cis-2-Butene-1,4-diol is a small organic diol used as a synthetic intermediate and as a probe in studies of isomerization, hydrogenation, and hydrogenolysis reactions. It is supplied for research use and is typically handled as a colorless to light-yellow liquid.

  • Purity 96.0%.
  • Molecular formula C4H8O2; molecular weight 88.11 g/mol.
  • Appearance colorless to light-yellow liquid; density 1.072 g/cm3.
  • Soluble in DMSO at 100 mg/mL; sonication recommended for dissolution.
  • Commonly used as an intermediate and mechanistic probe in synthetic studies.
  • Storage: pure form -20°C (up to 3 years) or 4°C (up to 2 years).

Catalog No. 50-003-01561 Supplier Medchemexpress LLC Supplier No. HYW027631500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

cis-2-Butene-1,4-diol is a small organic diol used as a synthetic intermediate and as a probe in studies of isomerization, hydrogenation, and hydrogenolysis reactions. It is supplied for research use and is typically handled as a colorless to light-yellow liquid.

  • Purity 96.0%.
  • Molecular formula C4H8O2; molecular weight 88.11 g/mol.
  • Appearance colorless to light-yellow liquid; density 1.072 g/cm3.
  • Soluble in DMSO at 100 mg/mL; sonication recommended for dissolution.
  • Commonly used as an intermediate and mechanistic probe in synthetic studies.
  • Storage: pure form -20°C (up to 3 years) or 4°C (up to 2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.