Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-2-Decene 95.0+%, TCI America™
SDP

Catalog No. D21611ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21611ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D21611ML Supplier TCI America Supplier No. D21611ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20348-51-0
Molecular Formula C10H20
Molecular Weight (g/mol) 140.27
MDL Number MFCD00048946
InChI Key YKNMBTZOEVIJCM-HWKANZROSA-N
PubChem CID 5364559
IUPAC Name (E)-dec-2-ene
SMILES CCCCCCCC=CC

Specifications

Color Colorless
Boiling Point 61°C
Quantity 1 mL
UN Number 3295
Formula Weight 140.27
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-2-Decene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.