Learn More
Cayman Chemical cis-2-Decenoic Acid, Purity: ≥95%, CAS: 15790-91-7, 100 mg
SDP
Catalog No. 502955121
Encompass_Preferred
An unsaturated short chain fatty acid that is secreted by P. aeruginosa and induces a dispersion response in biofilms formed by gram-negative and gram-positive bacteria, as well as by the yeast C. albicans; promotes dispersion of P. aeruginosa biofilms over a concentration range of 1.0 to 10 nM
Catalog No. 50-295-5121 Supplier Cayman Chemical Supplier No. 11966100 MG
An unsaturated short chain fatty acid that is secreted by P. aeruginosa and induces a dispersion response in biofilms formed by gram-negative and gram-positive bacteria, as well as by the yeast C. albicans; promotes dispersion of P. aeruginosa biofilms over a concentration range of 1.0 to 10 nM