Learn More
Medchemexpress LLC cis-2-decenoic acid | 15790-91-7 | MFCD20543109 | 99.6% | 170.25 g/mol | C10H18O2 | 5 MG
SDP
Catalog No. 5000262367
Encompass_Preferred
(Z)-2-Decenoic acid is a cis-unsaturated fatty acid produced by Pseudomonas aeruginosa that induces dispersion of bacterial biofilms and inhibits biofilm development. It is provided as a high-purity research reagent suitable for in vitro studies and formulation development.
  • Induces dispersion in gram-negative and gram-positive biofilms.
  • High purity (99.56%) for reproducible experimental results.
  • Highly soluble in DMSO (≥ 100 mg/mL) for convenient stock solutions.
  • Multiple small-scale pack sizes available for research use.
  • Stable when stored at -20°C in pure form for long-term storage.
Catalog No. 50-002-62367 Supplier Medchemexpress LLC Supplier No. HY132125MG
(Z)-2-Decenoic acid is a cis-unsaturated fatty acid produced by Pseudomonas aeruginosa that induces dispersion of bacterial biofilms and inhibits biofilm development. It is provided as a high-purity research reagent suitable for in vitro studies and formulation development.
  • Induces dispersion in gram-negative and gram-positive biofilms.
  • High purity (99.56%) for reproducible experimental results.
  • Highly soluble in DMSO (≥ 100 mg/mL) for convenient stock solutions.
  • Multiple small-scale pack sizes available for research use.
  • Stable when stored at -20°C in pure form for long-term storage.