Learn More
Cayman Chemical cIs-2-DecenoIc AcId 50mg
SDP
Catalog No. 502954625
Encompass_Preferred
An unsaturated short chain fatty acid that is secreted by P. aeruginosa and induces a dispersion response in biofilms formed by gram-negative and gram-positive bacteria, as well as by the yeast C. albicans; promotes dispersion of P. aeruginosa biofilms over a concentration range of 1.0 to 10 nM
Catalog No. 50-295-4625 Supplier Cayman Chemical Supplier No. 1196650 MG
An unsaturated short chain fatty acid that is secreted by P. aeruginosa and induces a dispersion response in biofilms formed by gram-negative and gram-positive bacteria, as well as by the yeast C. albicans; promotes dispersion of P. aeruginosa biofilms over a concentration range of 1.0 to 10 nM