Learn More
Medchemexpress LLC Cis-2-(diphenylmethyl)-N-[(2-iodophenyl)methyl]-1-azabicyclo[2.2.2]octan-3-amine oxalate | MFCD11045930 | ≥98.0% | 598.47 g/mol | C29H31IN2O4 | 5 MG
SDP
Catalog No. 5000264424
Encompass_Preferred
L-703606 oxalate hydrate is the oxalate salt hydrate form of L-703606, a potent and selective antagonist of the neurokinin-1 (NK1) receptor. It is supplied as a solid research reagent for biochemical and pharmacological studies, including investigations of gastric acid secretion and NK1-mediated signaling. For research use only.
  • Potent, selective NK1 receptor antagonist.
  • Supplied as an oxalate salt hydrate solid.
  • High purity suitable for biochemical assays.
  • Useful for studies of gastric acid secretion and NK1 signaling.
  • Provided with batch-specific certificate of analysis.
Catalog No. 50-002-64424 Supplier Medchemexpress LLC Supplier No. HY117216B5MG
L-703606 oxalate hydrate is the oxalate salt hydrate form of L-703606, a potent and selective antagonist of the neurokinin-1 (NK1) receptor. It is supplied as a solid research reagent for biochemical and pharmacological studies, including investigations of gastric acid secretion and NK1-mediated signaling. For research use only.
  • Potent, selective NK1 receptor antagonist.
  • Supplied as an oxalate salt hydrate solid.
  • High purity suitable for biochemical assays.
  • Useful for studies of gastric acid secretion and NK1 signaling.
  • Provided with batch-specific certificate of analysis.