Learn More
AdipoGen cis-2-Dodecenoic acid
SDP
Catalog No. 502561144
Encompass_Preferred
Chemical. CAS 55928-65-9. Formula C12H22O2. MW 198.3. Synthetic. Burkholderia cenocepacia produces a diffusible fatty acid signal molecule, cis-2-dodecenoic acid BDSF, that has been implicated in interspecies and interkingdom communication.
Catalog No. 50-256-1144 Supplier AdipoGen Supplier No. CDXD0293M250
Chemical. CAS 55928-65-9. Formula C12H22O2. MW 198.3. Synthetic. Burkholderia cenocepacia produces a diffusible fatty acid signal molecule, cis-2-dodecenoic acid BDSF, that has been implicated in interspecies and interkingdom communication.