Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC cis-2-Hexen-1-ol | 928-94-9 | 91.3% | 100.16 | 10 G
SDP

Catalog No. 5000364960
Encompass_Preferred

cis-2-Hexen-1-ol is a biochemical reagent suitable for life science research. It serves as a biological material or organic compound and is intended for research use only, not for patient administration.

  • Appears as a colorless to light yellow liquid.
  • Chemical formula is C6H12O.
  • Ships at room temperature in the continental US.
  • Store pure form at -20°C for up to 3 years.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month.
  • Soluble in DMSO at 100 mg/mL.

Catalog No. 50-003-64960 Supplier Medchemexpress LLC Supplier No. 10GHYW127618
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

cis-2-Hexen-1-ol is a biochemical reagent suitable for life science research. It serves as a biological material or organic compound and is intended for research use only, not for patient administration.

  • Appears as a colorless to light yellow liquid.
  • Chemical formula is C6H12O.
  • Ships at room temperature in the continental US.
  • Store pure form at -20°C for up to 3 years.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month.
  • Soluble in DMSO at 100 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.