Learn More
Description
- Cis-2-hexene is one of the isomeric forms of hexene
- It is obtained from 2-hexyne, via semihydrogenation in the presence of pd78(phen) [phen = 1,10-phenanthroline] on titanium dioxide in n-octane
- Cis-2-hexene is formed as one of the primary product from the dimerization of propylene in the presence of a nickel oxide-silica-alumina catalyst
- It undergoes isomerization/hydrosilylation reaction with phenyl silane in the presence of cobalt catalyst to afford 1-hexylphenylsilane
Specifications
Specifications
| CAS | 7688-21-3 |
| Density | 0.669 g/mL (at 25°C (literature)) |
| Boiling Point | 68°C to 70°C (lit.) |
| Molecular Formula | C6H12 |
| Refractive Index | n20/D 1.396 (literature) |
| Linear Formula | CH3CH2CH2CH=CHCH3 |
| MDL Number | MFCD00066520 |
| Quantity | 1 g |
| Molecular Weight (g/mol) | 84.16 |
| Percent Purity | 95% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.