Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-2-Methoxycinnamic Acid 98.0+%, TCI America™
SDP

Catalog No. M07615G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M07615G 5 g
M076125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M07615G Supplier TCI America Supplier No. M07615G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14737-91-8
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD00004375
InChI Key FEGVSPGUHMGGBO-SREVYHEPSA-N
Synonym cis-O-Methyl-o-coumaric Acid, cis-O-Methyl-o-cumaric Acid
PubChem CID 1622530
IUPAC Name (Z)-3-(2-methoxyphenyl)prop-2-enoic acid
SMILES COC1=CC=CC=C1C=CC(=O)O

Specifications

Melting Point 94°C
Color White-Yellow
Quantity 5 g
Formula Weight 178.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material cis-2-Methoxycinnamic Acid
RTECSNumber GD9720000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.