Learn More
TARGETMOL CHEMICALS INC CIS-22A 5MG
SDP
Catalog No. 502579947
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Cis-22a is a TRPV6 inhibitor (IC50 = 0.32 uM) which exhibits selectivity against related TRPV channels and calcium channels. cis-22a displays antiproliferative effects on T47D human breast cancer cells. purity: 98%
Catalog No. 50-257-9947 Supplier TARGETMOL CHEMICALS INC Supplier No. T412335MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Cis-22a is a TRPV6 inhibitor (IC50 = 0.32 uM) which exhibits selectivity against related TRPV channels and calcium channels. cis-22a displays antiproliferative effects on T47D human breast cancer cells. purity: 98%