Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-3-Aminocyclohexanecarboxylic Acid 96.0+%, TCI America™
SDP

Catalog No. A21275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21275G 5 g
A212725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21275G Supplier TCI America Supplier No. A21275G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 16636-51-4 | C7H13NO2 | 143.186 g/mol

Chemical Identifiers

CAS 16636-51-4
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD09753647
InChI Key CKTUXQBZPWBFDX-RITPCOANSA-N
PubChem CID 3082488
IUPAC Name (1R,3S)-3-aminocyclohexane-1-carboxylic acid
SMILES C1CC(CC(C1)N)C(=O)O

Specifications

Melting Point 278°C
Color White-Yellow
Quantity 5 g
Formula Weight 143.19
Percent Purity ≥96.0% (GC,T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material cis-3-Aminocyclohexanecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.