Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-cis-3-(Carbobenzoxyamino)cyclohexanecarboxylic Acid 95.0+%, TCI America™
SDP

Catalog No. C23971G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C23971G Supplier TCI America Supplier No. C23971G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Crystalline powder reagent with high purity for laboratory use.

Chemical Identifiers

CAS 952616-39-6
Molecular Formula C15H19NO4
Molecular Weight (g/mol) 277.32
MDL Number MFCD20259654,MFCD09910158
InChI Key LTEORMGXUMWZCU-UHFFFAOYNA-N
Synonym (+/-)-cis-3-(Cbz-amino)cyclohexanecarboxylic Acid
PubChem CID 51341837
IUPAC Name 3-{[(benzyloxy)carbonyl]amino}cyclohexane-1-carboxylic acid
SMILES OC(=O)C1CCCC(C1)NC(=O)OCC1=CC=CC=C1

Specifications

Color White
Quantity 1 g
Formula Weight 277.32
Percent Purity ≥95.0% (HPLC,T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material (+/-)-cis-3-(Carbobenzoxyamino)cyclohexanecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.