Learn More
Medchemexpress LLC Cis-3-hexen-1-ol | 928-96-1 | 99.9% | 100.16 | 1 ML
SDP
Catalog No. 5000452923
Encompass_Preferred
cis-3-Hexen-1-ol ((Z)-3-Hexen-1-ol) is a green grassy smelling compound found in many fresh fruits and vegetables with bitter taste enhancement. It is widely used as an added flavor in processed food to provide a fresh green quality. cis-3-Hexen-1-ol is an attractant to various insects.
  • Green grassy smelling compound
  • Found in many fresh fruits and vegetables
  • Used as an added flavor in processed food
  • Provides a fresh green quality
  • Attractant to various insects
Catalog No. 50-004-52923 Supplier Medchemexpress LLC Supplier No. HYW01060710MM/1ML
cis-3-Hexen-1-ol ((Z)-3-Hexen-1-ol) is a green grassy smelling compound found in many fresh fruits and vegetables with bitter taste enhancement. It is widely used as an added flavor in processed food to provide a fresh green quality. cis-3-Hexen-1-ol is an attractant to various insects.
  • Green grassy smelling compound
  • Found in many fresh fruits and vegetables
  • Used as an added flavor in processed food
  • Provides a fresh green quality
  • Attractant to various insects