Learn More
Medchemexpress LLC Cis-3-hexen-1-ol | 928-96-1 | 99.9% | 100.16 | 5 G
SDP
Catalog No. 5000451890
Encompass_Preferred
cis-3-Hexen-1-ol is a green grassy smelling compound found in many fresh fruits and vegetables with bitter taste enhancement. It is widely used as an added flavor in processed food to provide a fresh green quality, and it also acts as an attractant to various insects.
  • Green grassy smelling compound
  • Bitter taste enhancement
  • Widely used as an added flavor in processed food
  • Provides a fresh green quality
  • Acts as an attractant to various insects
  • Shows spatial differences in emission from different Nicotiana benthamiana leaves
Catalog No. 50-004-51890 Supplier Medchemexpress LLC Supplier No. HYW0106075G
cis-3-Hexen-1-ol is a green grassy smelling compound found in many fresh fruits and vegetables with bitter taste enhancement. It is widely used as an added flavor in processed food to provide a fresh green quality, and it also acts as an attractant to various insects.
  • Green grassy smelling compound
  • Bitter taste enhancement
  • Widely used as an added flavor in processed food
  • Provides a fresh green quality
  • Acts as an attractant to various insects
  • Shows spatial differences in emission from different Nicotiana benthamiana leaves