Learn More
Sigma Aldrich Fine Chemicals Biosciences cis-3-Hexen-1-ol natural 9100G
Catalog No. 502898327
Encompass
cis-3-Hexen-1-ol is one of the key volatile constituents of green leaf volatiles(GLV) that can act as an attractant to various insects. It is emitted by green plants when they are physically damaged.
Catalog No. 50-289-8327 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W256323100GK
cis-3-Hexen-1-ol is one of the key volatile constituents of green leaf volatiles(GLV) that can act as an attractant to various insects. It is emitted by green plants when they are physically damaged.