Learn More
Sigma Aldrich Fine Chemicals Biosciences cis 3 Hexen 1 ol natural 91KG
Catalog No. 502914171
Encompass
cis-3-Hexen-1-ol is one of the key volatile constituents of green leaf volatiles(GLV) that can act as an attractant to various insects. It is emitted by green plants when they are physically damaged.
Catalog No. 50-291-4171 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W2563231KGK
cis-3-Hexen-1-ol is one of the key volatile constituents of green leaf volatiles(GLV) that can act as an attractant to various insects. It is emitted by green plants when they are physically damaged.