Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-3-Hexen-1-yl 2-Nonynoate, TCI America™
SDP

Catalog No. N059725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N059725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N059725ML Supplier TCI America Supplier No. N059725ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 68480-29-5
Molecular Formula C15H24O2
Molecular Weight (g/mol) 236.355
MDL Number MFCD00036626
InChI Key PDPNRGJWKDIKKR-VURMDHGXSA-N
Synonym 2-Nonynoic Acid cis-3-Hexen-1-yl Ester
PubChem CID 5367676
IUPAC Name [(Z)-hex-3-enyl] non-2-ynoate
SMILES CCCCCCC#CC(=O)OCCC=CCC

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 236.36
Physical Form Liquid
Chemical Name or Material cis-3-Hexen-1-yl 2-Nonynoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.