Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-3-Hexen-1-yl Benzoate 98.0+%, TCI America™
SDP

Catalog No. B103925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B103925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B103925ML Supplier TCI America Supplier No. B103925ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25152-85-6
Molecular Formula C13H16O2
Molecular Weight (g/mol) 204.269
MDL Number MFCD00036526
InChI Key BCOXBEHFBZOJJZ-ARJAWSKDSA-N
Synonym Benzoic Acid cis-3-Hexen-1-yl Ester
PubChem CID 5367706
IUPAC Name [(Z)-hex-3-enyl] benzoate
SMILES CCC=CCCOC(=O)C1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 135°C
Quantity 25 mL
Formula Weight 204.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-3-Hexen-1-yl Benzoate
EINECSNumber (3)-1357
RTECSNumber DH1442500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.