Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-3-Hexen-1-yl Tiglate 97.0+%, TCI America™
SDP

Catalog No. T093425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T093425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T093425ML Supplier TCI America Supplier No. T093425ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67883-79-8
Molecular Formula C11H18O2
Molecular Weight (g/mol) 182.263
MDL Number MFCD00036528
InChI Key JNWQKXUWZWKUAY-BHHIIOOYSA-N
Synonym Tiglic Acid cis-3-Hexen-1-yl Ester
PubChem CID 16220109
IUPAC Name [(Z)-hex-3-enyl] (Z)-2-methylbut-2-enoate
SMILES CCC=CCCOC(=O)C(=CC)C

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 182.26
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-3-Hexen-1-yl Tiglate
RTECSNumber EM9253500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.