Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-3-Hexene, 97%

Catalog No. AAH5348506
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAH5348506

Supplier: Thermo Scientific Chemicals H5348506

Only null left
Add to Cart

CAS: 7642-09-3 | C6H12 | 84.16 g/mol

cis-3-Hexene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-3-Hexene is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7642-09-3
Molecular Weight (g/mol) 84.16
MDL Number MFCD00085300
InChI Key ZQDPJFUHLCOCRG-WAYWQWQTSA-N
Synonym cis-3-hexene, z-3-hexene, 3-hexene, z, z-hex-3-ene, 3z-hex-3-ene, 3-hexene, 3z, unii-50181bz6yt, 3z-3-heptene, 3z-3-hexene #, 3-hexene cis
PubChem CID 643783
IUPAC Name (Z)-hex-3-ene
SMILES CCC=CCC

Specifications

Density 0.681
Boiling Point 66°C to 68°C
Flash Point −25°C (−13°F)
Molecular Formula C6H12
Refractive Index 1.395
Quantity 5 g
UN Number UN3295
Beilstein 1718858
Formula Weight 84.16
Percent Purity 97%
Chemical Name or Material cis-3-Hexene
Show More Show Less
Hazard Category H225-H304
Hazard Statement GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.